missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13076-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
DBF4 | |
Polyclonal | |
Unconjugated | |
DBF4 | |
Activator of S phase kinase, ASK, CHIF, Chiffon homolog A, DBF4, DBF4 homolog (S. cerevisiae), DBF4A, Protein DBF4 homolog A, ZDBF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10926, 27214, 312046 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QZ41, Q9UBU7 | |
DBF4 | |
DBF4 Fusion Protein Ag3771 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |