missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DBI |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16893144
|
Proteintech
14490-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16883144
|
Proteintech
14490-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Diazepam Binding Inhibitor (DBI) is a highly conserved 10 kDa polypeptide which is expressed in various species range from yeast to mammals. As an inverse agonist for benzodiazepine receptors, DBI downregulates inhibitory effects of GABA. It also has potential to induce anxiety. Found in central and peripheral tissues, DBI also participates in metabolism of steroids, which has been known to partially modify GABAA receptor function in the CNS. In peripheral tissues, DBI plays regulatory roles in steroidogenesis. DBI levels have been reported to be decreased in the cerebrospinal fluid obtained from patients with Alzheimer disease.Specifications
DBI | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P07108, P11030, P31786 | |
13167, 1622, 25045 | |
DBI Fusion Protein Ag5891 | |
Primary | |
-20°C | |
DBI |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACBD1, ACBP, Acyl CoA binding protein, CCK RP, DBI, Diazepam binding inhibitor, Endozepine, EP | |
Dbi | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |