missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14490-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Diazepam Binding Inhibitor (DBI) is a highly conserved 10 kDa polypeptide which is expressed in various species range from yeast to mammals. As an inverse agonist for benzodiazepine receptors, DBI downregulates inhibitory effects of GABA. It also has potential to induce anxiety. Found in central and peripheral tissues, DBI also participates in metabolism of steroids, which has been known to partially modify GABAA receptor function in the CNS. In peripheral tissues, DBI plays regulatory roles in steroidogenesis. DBI levels have been reported to be decreased in the cerebrospinal fluid obtained from patients with Alzheimer disease.Specifications
DBI | |
Polyclonal | |
Unconjugated | |
DBI | |
ACBD1, ACBP, Acyl CoA binding protein, CCK RP, DBI, Diazepam binding inhibitor, Endozepine, EP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13167, 1622, 25045 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07108, P11030, P31786 | |
Dbi | |
DBI Fusion Protein Ag5891 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |