missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAMKL2 (DCK2) Recombinant Human Protein
DCAMKL2 (DCK2) Recombinant Human Protein
Brand: Invitrogen PV4297
Additional Details : Weight : 0.00900kg
Description
DCAMKL2 is a serine⁄threonine kinase involved in signal transduction and cellular communication, and is the human homolog of a rat protein kinase expressed in cerebrocortical lamination that binds microtubules.Specifications
NP_689832 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
DCAMKL2 (DCK2) Recombinant Human Protein | |
–80°C | |
1 Each | |
DCAMKL2 (DCK2) Recombinant Human Protein | |
DCAMKL kinase family | |
33nmoles of phosphate transferred to Autocamtide-2 peptide substrate (KKALRRQETVDAL) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinases and inhibitors | |
85% by SDS-PAGE |
0.40mg/mL | |
103.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
DCAMKL2 (DCK2) | |
DCAMKL2 was pre-diluted in enzyme dilution buffer and assayed in 10mM HEPES (pH 7.5), 10mM MgCl2, 1mM Na3VO4, 10μg/mL calmodulin, 1mM CaCl2, 2.5mM DTT, 200μM ATP, 100μM Autocamtide-2 peptide substrate (KKALRRQETVDAL) and trace [32P]-γ- ATP for 10 minutes at 30°C | |
DCAMKL2 (DCK2) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.