missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCC-Specific Mouse anti-Human, Unconjugated, Clone: 2G4G4, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DCC-Specific |
---|---|
Clone | 2G4G4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849715
|
Proteintech
67203-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839715
|
Proteintech
67203-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The DCC-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DCC-Specific. This antibody recognizes human, mouse antigen. The DCC-Specific antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
DCC-Specific | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1630 | |
Primary |
2G4G4 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG2a |