Learn More
DCC-Specific Mouse anti-Human, Unconjugated, Clone: 2G4G4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67203-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839715
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The DCC-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DCC-Specific. This antibody recognizes human, mouse antigen. The DCC-Specific antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
DCC-Specific
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Human
ELISA, Western Blot
2G4G4
Unconjugated
150 μL
1630
IgG2a