Läs mer
Tocris Bioscience™ DCEBIO
Beskrivning
DCEBIO is a stimulates Cl- secretion via the activation of hKCa3.1 (IK1) potassium channels and an apical Cl- conductance carried by the CFTR channel. More potent than its analog 1-EBIO (Cat. No. 1041).
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5,6-Dichloro-1-ethyl-1,3-dihydro-2H-benzimidazol-2-one |
| CAS | 60563-36-2 |
| Analysprocentintervall | >99% |
| Kvantitet | 50 mg |
| Löslighet | Soluble to 100mM in DMSO |
| Molekylformel | C9H8Cl2N2O |
| Formel vikt | Observed MW: 231.08 |
For Research Use Only