missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
DCK Mouse anti-Human, Clone: OTI16E12, lyophilized, TrueMAB™
Beskrivning
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Specifikationer
Specifikationer
| Antigen | DCK |
| Användningsområden | Flow Cytometry, Western Blot |
| Klassificering | Monoclonal |
| Klona | OTI16E12 |
| Konjugera | Unconjugated |
| Formulering | PBS with 8% trehalose and no preservative; pH 7.3 |
| Gen | DCK |
| Genaccessionsnr. | P27707 |
| Gene Alias | deoxycytidine kinase |
| Gensymboler | DCK |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?