Learn More
DCPS Mouse anti-Human, Unconjugated, Clone: 1B2E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67311-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839865
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The DCPS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DCPS. This antibody recognizes human antigen. The DCPS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
DCPS
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunofluorescence, Immunohistochemistry, Western Blot
1B2E4
Unconjugated
150 μL
28960
IgG2b