missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ DDHD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100466
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Antibody detects endogenous levels of total DDHD2.
DDHD2, also known as KIAA0725p, is a member of the intracellular phospholipase A1 (PLA1) protein family which comprises a group of enzymes that hydrolyze the sn-1 ester bond of phospholipids, producing 2-acyl-lysophospholipids and fatty acids. DDHD2 prefers phosphatidic acid as substrate and has a role in efficient membrane trafficking from the Golgi apparatus to the plasma membrane. The gene of DDHD2 maps to chromosome 8p11.23, and encodes a 711-amino-acid protein with a calculated molecular mass of 81 kDa. It has been reported that DDHD2 immunoblot analysis of various tissue extracts revealed two bands of 90 and 85 kDa.
Specifikationer
| DDHD2 | |
| Polyclonal | |
| Unconjugated | |
| DDHD2 | |
| 2010305K11Rik; DDHD domain containing 2; DDHD domain-containing protein 2; Ddhd2; intracellular phospholipase A1gamma; iPLA(1)gamma; KIAA0725; KIAA0725p; mKIAA0725; PA-PLA1 like; phospholipase DDHD2; SAM, WWE and DDHD domain-containing protein 1; Samwd1; sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipase A1; SPG54 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23259, 680971, 72108 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94830, Q80Y98 | |
| DDHD2 | |
| A synthesized peptide derived from human DDHD2(Accession O94830), corresponding to amino acid residues M1-V40. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering