missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DDIT3 (Human) Recombinant Protein
Human DDIT3 full-length ORF ( AAH03637.1, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001649-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAAESLPFSFGTLSSWELEAWYEDLQEVLSSDENGGTYVSPPGNEEEESKIFTTLDPASLAWLTEEEPEPAEVTSTSQSPHSPDSSQSSLAQEEEEEDQGRTRKRKQSGHSPARAGKQRMKEKEQENERKVAQLAEENERLKQEIERLTREVEATRRALIDRMVNLHQA
Specifications
AAH03637.1 | |
DNA-damage-inducible transcript 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDIT3 | |
GST |
1649 | |
Wheat germ expression system | |
10 μg | |
CEBPZ, CHOP, CHOP10, GADD153, MGC4154 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |