Läs mer
Tocris Bioscience™ DDR1-IN-1 dihydrochloride
Selective DDR1 inhibitor
Brand: Tocris Bioscience™ 5077/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
DDR1-IN-1 dihydrochloride is a selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.Specifikationer
| >98% | |
| 1780303-76-5 | |
| 10 mg |
Produkttitel
For Research Use Only