missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11324-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Gemin 3 is a member of the DEAD box protein family which are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. DEAD box proteins such as Gemin 3 are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of DEAD box family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 has an ATPase activity and a component of the survival of motor neurons (SMN) complex. The Gemin 3 protein may play a catalytic role in the function of the SMN complex on RNPs. Diseases associated with Gemin 3 protein dysfunction include spinal muscular dystrophy and muscular atrophy.Specifications
DDX20 | |
Polyclonal | |
Unconjugated | |
DDX20 | |
Component of gems 3, DDX20, DEAD box protein 20, DEAD box protein DP 103, DP103, Gemin 3, GEMIN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11218, 53975, 84473 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JJY4, Q9UHI6 | |
Ddx20 | |
DDX20 Fusion Protein Ag1863 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |