Läs mer
DDX3-IN-1, MedChemExpress
Produktkod. 16471247
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16471247 50 mg -
16455008 10 mg -
16437737 5 mg 5mg
16435766 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16471247 Leverantör Medchem Express Leverantörsnummer HY121832
MedChemExpress DDX3-IN-1 (Compound 16f) is a DEAD-box polypeptide 3 (DDX3) inhibitor with CC50s of 50 and 36 μM for HIV and HCV, respectively. Antiviral activity.
DDX3-IN-1 (Compound 16f) is a DEAD-box polypeptide 3 (DDX3) inhibitor with CC50s of 50 and 36 μM for HIV and HCV, respectively. Antiviral activity.
Specifikationer
Kemiskt namn eller material DDX3-IN-1
Fysisk form Solid
CAS 1919828-83-3
Färg Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H17N5O
För användning med (applikation) COVID-19-anti-virus
Löslighetsinformation DMSO : 125 mg/mL (406.70 mM; Need ultrasonic)
LEDER O=C(NC1=CC=C(N2N=NC(C)=C2)C=C1)NC3=CC=CC=C3C
Molekylvikt (g/mol) 307.35
Formel vikt 307.35
Procent renhet 99.57%