missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX4 VASA Mouse anti-Human, Unconjugated, Clone: 1F12C3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DDX4 VASA |
---|---|
Clone | 1F12C3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849615
|
Proteintech
67147-2-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839615
|
Proteintech
67147-2-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The DDX4, VASA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX4, VASA. This antibody recognizes human, mouse antigen. The DDX4, VASA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
DDX4 VASA | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
54514 | |
Primary |
1F12C3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG1 |