missing translation for 'onlineSavingsMsg'
Learn More

DDX43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17591-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827524

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
Specifications

Specifications

DDX43
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NXZ2
Cancer/testis antigen 13, CT13, DDX43, DEAD box protein 43, DEAD box protein HAGE, HAGE, Helical antigen
Rabbit
IgG
150 μL
-20°C
Primary
100048658, 55510, 682138
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DDX43
Liquid
DDX43
DDX43 Fusion Protein Ag11484
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers