Läs mer
De-4-fluoro 5-Fluoro Chidamide, TRC
Produktkod. 30365608
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30365608 250 mg
1 options
Produktkod. 30365608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289910250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material De-4-fluoro 5-Fluoro Chidamide
CAS 743420-02-2
Molekylformel C22H19FN4O2
Kvantitet 250 mg
Synonym N-(2-Amino-4-fluorophenyl)-4-[[[1-oxo-3-(3-pyridinyl)-2-propenyl]amino]methyl]benzamide, Isochidamide, (E)-N-(2-Amino-4-fluorophenyl)-4-((3-(pyridin-3-yl)acrylamido)methyl)benzamide
InChI formel InChI=1S/C22H19FN4O2/c23-18-8-9-19(24)20(12-18)27-22(29)17-6-3-16(4-7-17)14-26-21(28)10-5-15-2-1-11-25-13-15/h1-13H,14,24H2,(H,26,28)(H,27,29)/b10-5+
LEDER O=C(NCC1=CC=C(C(NC2=CC(F)=CC=C2N)=O)C=C1)/C=C/C3=CC=CN=C3
Molekylvikt (g/mol) 390.41