Läs mer
De-boc Cabazitaxel, TRC
Produktkod. 30012760
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012760 5 mg
30003930 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2060005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1638286-64-2
Molekylformel C40H49NO12
Molekylvikt (g/mol) 735.82
Synonym Benzenepropanoic acid, β-amino-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)- (ACI), (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4,6-dimethoxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl (αR,βS)-β-amino-α-hydroxybenzenepropanoate (ACI)
IUPAC-namn [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-acetyloxy-15-[(2R,3S)-3-amino-2-hydroxy-3-phenylpropanoyl]oxy-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
LEDER CO[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](N)c6ccccc6)C(=C([C@@H](OC)C(=O)[C@]13C)C5(C)C)C
Specifikationer
Kemiskt namn eller material De-boc Cabazitaxel
Kvantitet 5 mg
InChI formel InChI=1S/C40H49NO12/c1-21-25(51-36(46)30(43)29(41)23-14-10-8-11-15-23)19-40(47)34(52-35(45)24-16-12-9-13-17-24)32-38(5,33(44)31(49-7)28(21)37(40,3)4)26(48-6)18-27-39(32,20-50-27)53-22(2)42/h8-17,25-27,29-32,34,43,47H,18-20,41H2,1-7H3/t25-,26-,27+,29-,30+,31+,32-,34-,38+,39-,40+/m0/s1