Läs mer
De(diethylaminomethyldiiodo) Amiodarone, TRC
Produktkod. 17934235
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17934235 1 g
17954775 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17934235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2284201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83790-87-8
Molekylformel C20 H20 O3
Molekylvikt (g/mol) 308.37
Synonym Methanone, (2-butyl-3-benzofuranyl)(4-methoxyphenyl)-, Ketone, 2-butyl-3-benzofuranyl p-methoxyphenyl (7CI), (2-Butyl-3-benzofuranyl)(4-methoxyphenyl)methanone, (2-Butylbenzofuran-3-yl)(4-methoxyphenyl)methanone
IUPAC-namn (2-butyl-1-benzofuran-3-yl)-(4-methoxyphenyl)methanone
LEDER CCCCc1oc2ccccc2c1C(=O)c3ccc(OC)cc3
Specifikationer
Kemiskt namn eller material De(diethylaminomethyldiiodo) Amiodarone
Kvantitet 1 g
InChI formel InChI=1S/C20H20O3/c1-3-4-8-18-19(16-7-5-6-9-17(16)23-18)20(21)14-10-12-15(22-2)13-11-14/h5-7,9-13H,3-4,8H2,1-2H3
Formel vikt 308.1412