Läs mer
De-O-ethoxy Tamsulosin, TRC
Produktkod. 30352259
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352259 2.5 g
30350041 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352259 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2284952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2244986-82-9
Molekylformel C18H24N2O4S
Molekylvikt (g/mol) 364.46
Synonym (R)-2-Methoxy-5-(2-((2-phenoxyethyl)amino)propyl)benzenesulfonamide
IUPAC-namn (R)-2-methoxy-5-(2-((2-phenoxyethyl)amino)propyl)benzenesulfonamide
LEDER O=S(C1=CC(C[C@H](NCCOC2=CC=CC=C2)C)=CC=C1OC)(N)=O
Specifikationer
Kemiskt namn eller material De-O-ethoxy Tamsulosin
Kvantitet 2.5 g
InChI formel InChI=1S/C18H24N2O4S/c1-14(20-10-11-24-16-6-4-3-5-7-16)12-15-8-9-17(23-2)18(13-15)25(19,21)22/h3-9,13-14,20H,10-12H2,1-2H3,(H2,19,21,22)/t14-/m1/s1
Formel vikt 364.15