Läs mer
Deacetyl Bisacodyl, TRC
Produktkod. 30011709
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011709 250 mg
30009171 500 mg
17939634 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288670250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 603-41-8
Molekylformel C18 H15 N O2
Molekylvikt (g/mol) 277.32
Synonym 4,4'-[(Pyridin-2-yl)methylene]diphenol, 4,4'-(Pyridin-2-ylmethylene)diphenol, Sodium Picosulfate Imp. B (EP), Bisacodyl Imp. A (EP), Bisacodyl-desacetyl
IUPAC-namn 4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenol
LEDER Oc1ccc(cc1)C(c2ccc(O)cc2)c3ccccn3
Specifikationer
Kemiskt namn eller material 4,4'-(Pyridin-2-ylmethylene)diphenol
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C18H15NO2/c20-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(21)11-7-14/h1-12,18,20-21H
Formel vikt 277.1103
Procent renhet >95%