Läs mer
Deacetyl Linezolid Thioacetamide, TRC
Produktkod. 30367719
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367719 500 mg
30367397 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367719 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD198970500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 216868-57-4
Molekylformel C16 H20 F N3 O3 S
Molekylvikt (g/mol) 353.41
Synonym Linezolid Related Compound B (USP)
IUPAC-namn N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]ethanethioamide
LEDER CC(=S)NC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
Specifikationer
Kemiskt namn eller material (S)-N-{[3-(3-fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl}thioacetamide
Kvantitet 500 mg
InChI formel InChI=1S/C16H20FN3O3S/c1-11(24)18-9-13-10-20(16(21)23-13)12-2-3-15(14(17)8-12)19-4-6-22-7-5-19/h2-3,8,13H,4-7,9-10H2,1H3,(H,18,24)/t13-/m0/s1
Formel vikt 353.1209