Läs mer
Deacetyl-O-demethyl Diltiazem Hydrobromide, TRC
Produktkod. 30364385
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364385 50 mg
30361842 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19893050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142926-10-1
Molekylformel C19H23BrN2O3S
Molekylvikt (g/mol) 439.37
Synonym (2S,3S)-5-[2-(Dimethylamino)ethyl]-2,3-dihydro-3-hydroxy-2-(4-hydroxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrobromide, Deacetyl-O-demethyl-d-diltiazem Hydrobromide, Deacetyl-O-demethyldiltiazem Hydrobromide
IUPAC-namn (2S,3S)-5-[2-(dimethylamino)ethyl]-3-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4-one;hydrobromide
LEDER Br.CN(C)CCN1C(=O)[C@H](O)[C@@H](Sc2ccccc12)c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material Deacetyl-O-demethyl Diltiazem Hydrobromide
Kvantitet 50 mg
InChI formel InChI=1S/C19H22N2O3S.BrH/c1-20(2)11-12-21-15-5-3-4-6-16(15)25-18(17(23)19(21)24)13-7-9-14(22)10-8-13;/h3-10,17-18,22-23H,11-12H2,1-2H3;1H/t17-,18+;/m1./s1
Formel vikt 438.0613