Läs mer
Deacetyl (R)-Linezolid, TRC
Produktkod. 30356061
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356061 1 g
30363074 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356061 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1988701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 912359-80-9
Molekylformel C14H18FN3O3
Molekylvikt (g/mol) 295.31
Synonym (5R)-5-(Aminomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
IUPAC-namn (5R)-5-(aminomethyl)-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one
LEDER NC[C@@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
Specifikationer
Kemiskt namn eller material Deacetyl (R)-Linezolid
Kvantitet 1 g
InChI formel InChI=1S/C14H18FN3O3/c15-12-7-10(18-9-11(8-16)21-14(18)19)1-2-13(12)17-3-5-20-6-4-17/h1-2,7,11H,3-6,8-9,16H2/t11-/m1/s1
Formel vikt 295.1332