Läs mer
Deacetyl Racecadotril, TRC
Produktkod. 30355596
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30355596 100 mg
1 options
Produktkod. 30355596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD198995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyl [[(2RS)-2-Benzyl-3-sulfanylpropanoyl]amino]acetate
CAS 81110-69-2
Molekylformel C19 H21 N O3 S
Kvantitet 100 mg
Synonym N-[2-(Mercaptomethyl)-1-oxo-3-phenylpropyl]glycine phenylmethyl ester, Benzyl [[(2RS)-2-Benzyl-3-sulfanylpropanoyl]amino]acetate
InChI formel InChI=1S/C19H21NO3S/c21-18(23-13-16-9-5-2-6-10-16)12-20-19(22)17(14-24)11-15-7-3-1-4-8-15/h1-10,17,24H,11-14H2,(H,20,22)
LEDER SCC(Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2
IUPAC-namn benzyl 2-[(2-benzyl-3-sulfanylpropanoyl)amino]acetate
Molekylvikt (g/mol) 343.44
Formel vikt 343.1242