Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Brand:  Trajan Scientific and Medical™ 06244501
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248803
  • 493.00 SEK / 2m
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifications
Specifications
Organic and Aqueous Solvents
1/Pk.
0.363 mm
Thermally stable up to 380°C
Tubing
0.125 mm
2 m