Läs mer
DEAE-Sephadex A-25 Chloride, TRC
Produktkod. 30354698
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30354698 2.5 g
1 options
Produktkod. 30354698 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1990602.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DEAE-Sephadex A-25 Chloride
CAS 12609-80-2
Kvantitet 2.5 g
Synonym DEAE-Sephadex A-25 Chloride