Läs mer
Debrisoquin-13C,15N2 Hemisulfate, TRC
Produktkod. 30350034
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350034 10 mg
30358708 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350034 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD20870210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246814-89-0
Molekylformel C913CH13N15N2•1/2H2SO4
Molekylvikt (g/mol) 227.25
Synonym 3,4-Dihydro-2(1H)-isoquinolinecarboximidamide-13C,15N2 Hemisulfate, Debrisoquine-13C,15N2 Hemisulfate, Isocaramidine-13C,15N2 Hemisulfate
IUPAC-namn 3,4-dihydro-1H-isoquinoline-2-(15N2)carboximidamide;sulfuric acid
LEDER [15NH]=[13C]([15NH2])N1CCC2=CC=CC=C2C1.O=S(O)(O)=O
Specifikationer
Kemiskt namn eller material Debrisoquin-13C,15N2 Hemisulfate
Kvantitet 10 mg
InChI formel InChI=1S/C10H13N3.H2O4S/c11-10(12)13-6-5-8-3-1-2-4-9(8)7-13;1-5(2,3)4/h1-4H,5-7H2,(H3,11,12);(H2,1,2,3,4)/i10+1,11+1,12+1;