Läs mer
Debrisoquin Hemisulfate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Debrisoquine Sulfate |
| CAS | 581-88-4 |
| Molekylformel | 2 C10 H13 N3 . H2 O4 S |
| Kvantitet | 500 mg |
| Synonym | 2(1H)-Isoquinolinecarboximidamide, 3,4-dihydro-, sulfate (2:1), 2(1H)-Isoquinolinecarboxamidine, 3,4-dihydro-, sulfate (2:1) (8CI), 1,2,3,4-Tetrahydro-isoquinoline-2-carboxamidine sulfate, Debrisoquin hemisulfate, Debrisoquin sulfate, Debrisoquine hemisulfate, Debrisoquine sulfate, Declinax, Isocaramidine sulfate, Ro 5-3307/1, Debrisoquin Sulphate |
| InChI formel | InChI=1S/2C10H13N3.H2O4S/c2*11-10(12)13-6-5-8-3-1-2-4-9(8)7-13;1-5(2,3)4/h2*1-4H,5-7H2,(H3,11,12);(H2,1,2,3,4) |
| LEDER | NC(=N)N1CCc2ccccc2C1.NC(=N)N3CCc4ccccc4C3.OS(=O)(=O)O |
| IUPAC-namn | 3,4-dihydro-1H-isoquinoline-2-carboximidamide;sulfuric acid |
| Molekylvikt (g/mol) | 448.54 |
| Formel vikt | 448.1893 |