missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
DEC1 Polyclonal antibody specifically detects DEC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ChIP assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, ChIP assay (ChIP), ChIP assay Sequencing
Specifikationer
Specifikationer
| Antigen | DEC1 |
| Användningsområden | Western Blot, Simple Western, ChIP Assay, Flow Cytometry, ELISA |
| Klassificering | Polyclonal |
| Konjugera | DyLight 755 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14 |
| Värd art | Rabbit |
| Immunogen | Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503] |
| Reningsmetod | Immunogen affinity purified |
| Kvantitet | 0.1 mL |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?