missing translation for 'onlineSavingsMsg'
Läs mer

DEC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Produktkod. 18726140 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
Förpackningsstorlek:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18726140 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18726140 Leverantör Novus Biologicals Leverantörsnummer NB1001800JF646

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

DEC1 Polyclonal antibody specifically detects DEC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ChIP assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, ChIP assay (ChIP), ChIP assay Sequencing
TRUSTED_SUSTAINABILITY

Spécification

Antigen DEC1
Användningsområden Western Blot, Simple Western, ChIP Assay, Flow Cytometry, ELISA
Klassificering Polyclonal
Konjugera Janelia Fluor 646
Formulering 50mM Sodium Borate
Gene Alias basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14
Värd art Rabbit
Immunogen Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503]
Reningsmetod Immunogen affinity purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin HIF Target Genes, Hypoxia
Primär eller sekundär Primary
Gen-ID (Entrez) 8553
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C in the dark.
Form Purified
Isotyp IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.