Läs mer
Decabromodiphenyl Ether-13C12, TRC
Produktkod. 30011833
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011833 5 mg
30011667 10 mg
30006959 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2128525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 562099-68-7
Molekylformel 13C12 Br10 O
Molekylvikt (g/mol) 971.08
Synonym Benzene-13C6, 1,1'-oxybis[2,3,4,5,6-pentabromo-, 1,1'-Oxybis[2,3,4,5,6-pentabromobenzene-13C6], BDE-209-13C12, PBDE 209 (13C12), Decabromodiphenyl ether (13C12), Decabromodiphenyl ether 13C12
IUPAC-namn 1,2,3,4,5-pentabromo-6-(2,3,4,5,6-pentabromo(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)oxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
LEDER Br[13c]1[13c](Br)[13c](Br)[13c](O[13c]2[13c](Br)[13c](Br)[13c](Br)[13c](Br)[13c]2Br)[13c](Br)[13c]1Br
Specifikationer
Kemiskt namn eller material Decabromodiphenyl Ether 13C12
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C12Br10O/c13-1-3(15)7(19)11(8(20)4(1)16)23-12-9(21)5(17)2(14)6(18)10(12)22/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1
Formel vikt 961.2185
Procent renhet >95%