Läs mer
Decabromodiphenylethane, TRC
Produktkod. 30008458
Change view
Kvantitet:
10 mg
100 mg
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008458 25 g
17990705 10 mg
17984355 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008458 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD21280025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84852-53-9
Molekylformel C14 H4 Br10
Molekylvikt (g/mol) 971.22
Synonym 1,1'-(1,2-Ethanediyl)bis[2,3,4,5,6-pentabromobenzene], 1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane, 1,2-Bis(pentabromophenyl)ethane, Bis(pentabromophenyl)ethane, CG 801, Decabromodiphenylethane, Decadiphenyl 8010, Ethylenebis(pentabromobiphenyl), Ethylenebispentabromobenzene, F 8010, FCP 801, FR 105, FR 1410, Firemaster 2100, Planelon BDE, RDT 3, RDT 3K, S 8010, Saytex 4010, Saytex 8010, Yabao 8010
IUPAC-namn 1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
LEDER Brc1c(Br)c(Br)c(CCc2c(Br)c(Br)c(Br)c(Br)c2Br)c(Br)c1Br
Specifikationer
Kemiskt namn eller material 1,2-Bis(pentabromophenyl)ethane
Kvantitet 25 g
InChI formel InChI=1S/C14H4Br10/c15-5-3(6(16)10(20)13(23)9(5)19)1-2-4-7(17)11(21)14(24)12(22)8(4)18/h1-2H2
Formel vikt 961.2147