missing translation for 'onlineSavingsMsg'
Learn More

Decafluorobenzophenone, 97%, Thermo Scientific™

Catalog Number p-7077162
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
853-39-4
C13F10O
362.13
WWQLXRAKBJVNCC-UHFFFAOYSA-N
decafluorobenzophenone, benzophenone, decafluoro, perfluorobenzophenone, methanone, bis pentafluorophenyl, bis perfluorophenyl methanone, bis pentafluorophenyl methanone, bis 2,3,4,5,6-pentafluorophenyl methanone, di2,3,4,5,6-pentafluorophenyl ketone, acmc-209q5n
70068
bis(2,3,4,5,6-pentafluorophenyl)methanone
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F
This item is not returnable. View return policy
853-39-4
C13F10O
362.13
WWQLXRAKBJVNCC-UHFFFAOYSA-N
decafluorobenzophenone, benzophenone, decafluoro, perfluorobenzophenone, methanone, bis pentafluorophenyl, bis perfluorophenyl methanone, bis pentafluorophenyl methanone, bis 2,3,4,5,6-pentafluorophenyl methanone, di2,3,4,5,6-pentafluorophenyl ketone, acmc-209q5n
70068
bis(2,3,4,5,6-pentafluorophenyl)methanone
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F
Melting Point 91.0°C to 95.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6F5)2CO
Quantity 1 g
Formula Weight 362.13
Percent Purity 97%
Chemical Name or Material Decafluorobenzophenone
compliance-icons
Product Identifier
  • Decafluorobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'