Läs mer
Decamethyl-1,9-Pentasiloxanediol, TRC
Produktkod. 30349543
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30349543 50 mg
1 options
Produktkod. 30349543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD21246050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Decamethyl-1,9-Pentasiloxanediol
CAS 7445-36-5
Molekylformel C10H32O6Si5
Kvantitet 50 mg
Synonym 1,1,3,3,5,5,7,7,9,9-Decamethyl-1,9-pentasiloxanediol, 1,9-Dihydroxydecamethylpentasiloxane, Decamethylpentasiloxane-1,9-diol, Decamethylpentasiloxanediol
InChI formel InChI=1S/C10H32O6Si5/c1-17(2,11)13-19(5,6)15-21(9,10)16-20(7,8)14-18(3,4)12/h11-12H,1-10H3
LEDER C[Si](C)(O)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O
IUPAC-namn bis[[[hydroxy(dimethyl)silyl]oxy-dimethylsilyl]oxy]-dimethylsilane
Molekylvikt (g/mol) 388.78