Läs mer
Decanoyl-L-carnitine Chloride, TRC
Produktkod. 30132820
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30132820 2.5 g 2.5g
30008177 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2125802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 369651-88-7
Molekylformel C17H34ClNO4
Molekylvikt (g/mol) 351.91
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium Chloride
IUPAC-namn (3R)-3-decanoyloxy-4-(trimethylazaniumyl)butanoate;hydrochloride
LEDER CC(=O)O[C@@H]1[C@@H](N2CCCCC2)C[C@]3(C)[C@@H]4[C@@H](CC[C@H]3C1)[C@H]5[C@@](CC4)(C)[C@@H](O)[C@@H](N6CCCCC6)C5
Specifikationer
Kemiskt namn eller material Decanoyl-L-carnitine Chloride
Kvantitet 2.5 g
InChI formel InChI=1S/C17H33NO4.ClH/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4;/h15H,5-14H2,1-4H3;1H/t15-;/m1./s1