missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Decanoylcarnitine Chloride, TRC
Specifications
Specifications
| Kemiskt namn eller material | (+/-)-Decanoylcarnitine Chloride |
| CAS | 18822-87-2 |
| Molekylformel | C17 H34 N O4 . Cl |
| Kvantitet | 100 mg |
| Synonym | Decanoyl Carnitine Hydrochloride, Decanoyl-d,l-carnitine Hydrochloride, Decanoylcarnitine Chloride, 3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium Chloride, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride (9CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride, (±)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, decanoate, (±)- (8CI), Decanoic acid, ester with (3-carboxy-2-hydroxypropyl)trimethylammonium chloride, (±)- |
| InChI formel | InChI=1S/C17H33NO4.ClH/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4;/h15H,5-14H2,1-4H3;1H |
| LEDER | [Cl-].CCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C |
| IUPAC-namn | (3-carboxy-2-decanoyloxy-propyl)-trimethyl-ammonium;chloride |
| Molekylvikt (g/mol) | 351.91 |
| Formel vikt | 351.2176 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?