Läs mer
(+/-)-Decanoylcarnitine Chloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (+/-)-Decanoylcarnitine Chloride |
| CAS | 18822-87-2 |
| Molekylformel | C17 H34 N O4 . Cl |
| Kvantitet | 25 mg |
| Synonym | Decanoyl Carnitine Hydrochloride, Decanoyl-d,l-carnitine Hydrochloride, Decanoylcarnitine Chloride, 3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium Chloride, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride (9CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-, chloride, (±)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, decanoate, (±)- (8CI), Decanoic acid, ester with (3-carboxy-2-hydroxypropyl)trimethylammonium chloride, (±)- |
| InChI formel | InChI=1S/C17H33NO4.ClH/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4;/h15H,5-14H2,1-4H3;1H |
| LEDER | [Cl-].CCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C |
| IUPAC-namn | (3-carboxy-2-decanoyloxy-propyl)-trimethyl-ammonium;chloride |
| Molekylvikt (g/mol) | 351.91 |
| Formel vikt | 351.2176 |