Läs mer
Decarbonyl Ropinirole Dihydrochloride, TRC
Produktkod. 30354466
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30354466 25 mg
1 options
Produktkod. 30354466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD21350025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Decarbonyl Ropinirole Dihydrochloride
CAS 1346603-36-8
Molekylformel C15 H26 N2 O
Kvantitet 25 mg
Synonym 2-Amino-6-(2-dipropylaminoethyl)phenyl)methanol Dihydrochloride
InChI formel InChI=1S/C15H26N2O/c1-3-9-17(10-4-2)11-8-13-6-5-7-15(16)14(13)12-18/h5-7,18H,3-4,8-12,16H2,1-2H3
LEDER CCCN(CCC)CCc1cccc(N)c1CO
IUPAC-namn [2-amino-6-[2-(dipropylamino)ethyl]phenyl]methanol
Molekylvikt (g/mol) 250.38
Formel vikt 250.205