Läs mer
Decarbonyl Zolmitriptan Dihydrochloride, TRC
Produktkod. 30354928
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30354928 100 mg
1 options
Produktkod. 30354928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD212960100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Decarbonyl Zolmitriptan Dihydrochloride
CAS 1241387-63-2
Molekylformel C15H25Cl2N3O
Kvantitet 100 mg
Synonym (betaS)-beta-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol Diydrochloride, (S)-2-Amino-3-[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]propan-1-ol Dihydrochloride
InChI formel InChI=1S/C15H23N3O.2ClH/c1-18(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13(16)10-19;;/h3-4,8-9,13,17,19H,5-7,10,16H2,1-2H3;2*1H/t13-;;/m0../s1
LEDER Cl.Cl.CN(C)CCc1c[nH]c2ccc(C[C@H](N)CO)cc12
IUPAC-namn (2S)-2-amino-3-[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]propan-1-ol;dihydrochloride
Molekylvikt (g/mol) 334.28
Formel vikt 333.1375