Läs mer
Decarboxy Etodolac, TRC
Produktkod. 30358122
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358122 100 mg
30365424 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358122 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD215350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115066-03-0
Molekylformel C16 H21 N O
Molekylvikt (g/mol) 243.34
Synonym (1RS)-1,8-Diethyl-1-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indole, Decarboxy Etodolac, Pyrano[3,4-b]indole, 1,8-diethyl-1,3,4,9-tetrahydro-1-methyl-, 1,8-Diethyl-1,3,4,9-tetrahydro-1-methylpyrano[3,4-b]indole, RAK 802
IUPAC-namn 1,8-diethyl-1-methyl-4,9-dihydro-3H-pyrano[3,4-b]indole
LEDER CCc1cccc2c3CCOC(C)(CC)c3[nH]c12
Specifikationer
Kemiskt namn eller material (1RS)-1,8-Diethyl-1-methyl-1,3,4,9-tetrahydropyrano(3,4-b)indole (Decarboxy Etodolac)
Kvantitet 100 mg
InChI formel InChI=1S/C16H21NO/c1-4-11-7-6-8-12-13-9-10-18-16(3,5-2)15(13)17-14(11)12/h6-8,17H,4-5,9-10H2,1-3H3
Formel vikt 243.1623