Läs mer
Dechlorane 603, TRC
Produktkod. 30134487
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134487 50 mg
30133505 100 mg
30041374 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134487 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22620550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13560-92-4
Molekylformel C17 H8 Cl12
Molekylvikt (g/mol) 637.68
Synonym 1,4:5,8:9,10-Trimethanoanthracene, 1,2,3,4,5,6,7,8,12,12,13,13-dodecachloro-1,4,4a,5,8,8a,9,9a,10,10a-decahydro-, 1,2,3,4,5,6,7,8,12,12,13,13-Dodecachloro-1,4,4a,5,8,8a,9,9a,10,10a-decahydro-1,4:5,8:9,10-trimethanoanthracene, Dec 603, Dechlorane 603
IUPAC-namn rac-(3R,6S,10R,13S)-3,4,5,6,10,11,12,13,16,16,17,17-dodecachlorohexacyclo[6.6.1.13,6.110,13.02,7.09,14]heptadeca-4,11-diene
LEDER ClC1=C(Cl)[C@@]2(Cl)C3[C@@H]4C[C@H](C3[C@]1(Cl)C2(Cl)Cl)C5C4[C@@]6(Cl)C(=C(Cl)[C@]5(Cl)C6(Cl)Cl)Cl
Specifikationer
Kemiskt namn eller material Dechlorane 603
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C17H8Cl12/c18-8-10(20)14(24)6-3-1-2(4(6)12(8,22)16(14,26)27)5-7(3)15(25)11(21)9(19)13(5,23)17(15,28)29/h2-7H,1H2/t2-,3+,4?,5?,6?,7?,12-,13+,14+,15-
Formel vikt 631.6888
Procent renhet >95%