Läs mer
Dechloro Haloperidol Decanoate, TRC
Produktkod. 30011742
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011742 100 mg
30012393 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD226510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797824-64-6
Molekylformel C31H42FNO3
Molekylvikt (g/mol) 495.67
Synonym 1-(4-(4-Fluorophenyl)-4-oxobutyl)-4-phenylpiperidin-4-yl Decanoate
IUPAC-namn 1-(4-(4-fluorophenyl)-4-oxobutyl)-4-phenylpiperidin-4-yl decanoate
LEDER O=C(C1=CC=C(C=C1)F)CCCN(CC2)CCC2(C3=CC=CC=C3)OC(CCCCCCCCC)=O
Specifikationer
Kemiskt namn eller material Dechloro Haloperidol Decanoate
Kvantitet 100 mg
InChI formel InChI=1S/C31H42FNO3/c1-2-3-4-5-6-7-11-16-30(35)36-31(27-13-9-8-10-14-27)21-24-33(25-22-31)23-12-15-29(34)26-17-19-28(32)20-18-26/h8-10,13-14,17-20H,2-7,11-12,15-16,21-25H2,1H3
Formel vikt 495.31