Läs mer
Decyl Methanethiosulfonate, TRC
Produktkod. 30363802
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30363802 50 mg
1 options
Produktkod. 30363802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22845050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Decyl Methanethiosulfonate
CAS 190852-38-1
Molekylformel C11 H24 O2 S2
Kvantitet 50 mg
Synonym Methanesulfonothioic acid, S-decyl ester, Decyl Methanethiosulfonate
InChI formel InChI=1S/C11H24O2S2/c1-3-4-5-6-7-8-9-10-11-14-15(2,12)13/h3-11H2,1-2H3
LEDER CCCCCCCCCCSS(=O)(=O)C
IUPAC-namn 1-methylsulfonylsulfanyldecane
Molekylvikt (g/mol) 252.44
Formel vikt 252.1218