Läs mer
Dediaminosulfonyl Hydroxymethyl Famotidine, TRC
Produktkod. 30366177
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30366177 250 mg
1 options
Produktkod. 30366177 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD227200250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dediaminosulfonyl Hydroxymethyl Famotidine
CAS 76823-94-4
Molekylformel C9H15N5OS2
Kvantitet 250 mg
Synonym 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-propanimidic Acid Methyl Ester, Methyl 3-(((2-((Diaminomethylene)amino)thiazol-4-yl)methyl)thio)propanimidate
InChI formel InChI=1S/C9H15N5OS2/c1-15-7(10)2-3-16-4-6-5-17-9(13-6)14-8(11)12/h5,10H,2-4H2,1H3,(H4,11,12,13,14)
LEDER COC(=N)CCSCc1csc(N=C(N)N)n1
IUPAC-namn methyl 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidate
Molekylvikt (g/mol) 273.37
Formel vikt 273.0718