Learn More
Scientific Industries SI™ Deepwell Microplate Foam Insert
Accommodates one standard footprint deepwell microplate. Scientific Industries SI™ Deepwell Microplate Foam Insert fits into the 152mm Platform Attachment.
Brand:  Scientific Industries SI™ 504-0232-00
Additional Details : Weight : 0.75000kg
Product Code. 15557325
  • 725.00 SEK / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • 3cm deep
  • Fits into152mm Platform attachment
  • For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Specifications
Specifications
High-density foam
3m deepwell microplates