Läs mer
OHAUS™ Defender™ 3000 Stainless Steel
Handla allt Ohaus Produkter
Kapacitet:
60 kg
150 kg
Modell:
D32XW150VL-EU
D32XW60VL-EU
D32XW60VR-EU
Förpackningsstorlek:
Varje
Beskrivning
- The NEMA 4X/IP66 stainless steel indicator is built to endure tough industrial elements.
- The 58-hour rechargeable battery offers continuous operation for multi-day operations.
- Durable construction with a 304 stainless steel platform, stainless steel tubular frame, column and indicator bracket, and IP67 stainless steel load cell.
- Easy-to-read display and basic counting mode allow for simple operation for everyday basic industrial applications.
- Offered with NTEP or OIML/EC Type approvals (depending on region)
Specifikationer
Specifikationer
| Batteritid | 58 Hours with Rechargeable Battery |
| Anslutningstyp | Easy access communication port with RS232 interface (included) |
| Konstruktion | IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, stainless steel column and wall bracket, 304 grade stainless steel platform with steel tubular frame, IP67 stainless steel load cell |
| Kapacitet | 60 kg |
| Visningstyp | Backlit LCD |
| Läsbarhet | 10.000000 g |
| Modell | D32XW60VL-EU |
| Skyddsklass | IP 65, NEMA 4X, IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, IP67 stainless steel load cell |
| Temperatur (metrisk) Drift | -10°C to +40°C |
| Vägningslägen | Gram, Kilogram, Ounce, Pound, Pound:Ounce |
| Visa mer |