Läs mer
Deferasirox Ethyl Ester, TRC
Produktkod. 30360550
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30360550 250 mg
1 options
Produktkod. 30360550 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD228655250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deferasirox Ethyl Ester
CAS 201530-79-2
Molekylformel C23 H19 N3 O4
Kvantitet 250 mg
Synonym Benzoic acid, 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-, ethyl ester, Ethyl 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoate, Deferasirox Ethyl Ester
InChI formel InChI=1S/C23H19N3O4/c1-2-30-23(29)15-11-13-16(14-12-15)26-22(18-8-4-6-10-20(18)28)24-21(25-26)17-7-3-5-9-19(17)27/h3-14,27-28H,2H2,1H3
LEDER CCOC(=O)c1ccc(cc1)n2nc(nc2c3ccccc3O)c4ccccc4O
IUPAC-namn ethyl 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoate
Molekylvikt (g/mol) 401.41
Formel vikt 401.1376