missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deferasirox Ethyl Ester, TRC
Specifications
Specifications
| Kemiskt namn eller material | Deferasirox Ethyl Ester |
| CAS | 201530-79-2 |
| Molekylformel | C23 H19 N3 O4 |
| Kvantitet | 100 mg |
| Synonym | Benzoic acid, 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-, ethyl ester, Ethyl 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoate, Deferasirox Ethyl Ester |
| InChI formel | InChI=1S/C23H19N3O4/c1-2-30-23(29)15-11-13-16(14-12-15)26-22(18-8-4-6-10-20(18)28)24-21(25-26)17-7-3-5-9-19(17)27/h3-14,27-28H,2H2,1H3 |
| LEDER | CCOC(=O)c1ccc(cc1)n2nc(nc2c3ccccc3O)c4ccccc4O |
| IUPAC-namn | ethyl 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoate |
| Molekylvikt (g/mol) | 401.41 |
| Formel vikt | 401.1376 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?