Läs mer
Deferasirox Ferrate(III) Tripotassium Complex Methanoate Hydrate, TRC
Produktkod. 30366912
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30366912 100 mg
1 options
Produktkod. 30366912 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD228670100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deferasirox Ferrate(III) Tripotassium Complex Methanoate Hydrate
CAS 554445-58-8
Molekylformel C44H36FeK3N6O12
Kvantitet 100 mg
Synonym (OC-6-22')-Bis[4-[3,5-bis[2-(hydroxy-kO)phenyl]-1H-1,2,4-triazol-1-yl-kN4]benzoato(3-)]ferrate(3-) Tripotassium Methanoate Hydrate
InChI formel InChI=1S/2C21H15N3O4.Fe/c2*25-17-7-3-1-5-15(17)19-22-20(16-6-2-4-8-18(16)26)24(23-19)14-11-9-13(10-12-14)21(27)28;/h2*1-12,25-26H,(H,27,28);/q;;+4/p-6
LEDER CO.CO.C1=C/C(=C/2\NN(/C(=C\3/C(=O)C=CC=C3)/N2)C4=CC=C(C=C4)C(=O)[O-])/C(=O)C=C1.C1=C/C(=C/2\NN(/C(=C\3/C(=O)C=CC=C3)/N2)C4=CC=C(C=C4)C(=O)[O-])/C(=O)C=C1.O.O.[K+].[K+].[K+].[Fe]
IUPAC-namn 4-[3,5-bis(2-oxidophenyl)-1,2,4-triazol-1-yl]benzoate;iron(4+)
Molekylvikt (g/mol) 1013.93